Triisopropanolamine, 95%
Triisopropanolamine, 95%
Triisopropanolamine, 95%
Thermo Scientific Chemicals

Triisopropanolamine, 95%

CAS: 122-20-3 | C9H21NO3 | 191.271 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10,000 g
500 g
2500 g
Catalog number A19966.0E
also known as A19966-0E
Price (EUR)
67,30
Each
Quantity:
2500 g
Request bulk or custom format
Price (EUR)
67,30
Each
Chemical Identifiers
CAS122-20-3
IUPAC Name1-[bis(2-hydroxypropyl)amino]propan-2-ol
Molecular FormulaC9H21NO3
InChI KeySLINHMUFWFWBMU-UHFFFAOYNA-N
SMILESCC(O)CN(CC(C)O)CC(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
FormFused solid or clear liquid as melt
Assay (Aqueous acid-base Titration)≥94.0 to ≤106.0%
Identification (FTIR)Conforms
Major applications include water-based coating applications and agricultural products. Additional applications are antistat agents for polymers, corrosion inhibitor, electrodeposition/electrocoating, lubricants, paper, pigment dispersion, plastics, polyurethane additive, reaction intermediates, rubber curing, surfactants, mineral dispersion, and urethanes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Major applications include water-based coating applications and agricultural products. Additional applications are antistat agents for polymers, corrosion inhibitor, electrodeposition/electrocoating, lubricants, paper, pigment dispersion, plastics, polyurethane additive, reaction intermediates, rubber curing, surfactants, mineral dispersion, and urethanes.

Solubility
Insoluble in water. Soluble ethanol, diethyl ether, chloroform (slightly ), and methanol (>500 g/100g).

Notes
Hygroscopic. Store away from oxidizing agents, water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
RUO – Research Use Only

General References:

  1. Pensri Piboonchaisit, et al. Novel Route to Tris(silatranyloxy-i-propyl)amine Directly from Silica and Triisopropanolamine, Part I.ScienceAsia.1999,25113-119.
  2. Paul J. Sandberg.; F. Doncaster. On the mechanism of strength enhancement of cement paste and mortar with triisopropanolamine.Cem. Concr. Res.2004,34(6), 973-976.