Qubit™ Flex Fluorometers and Starter Kits
Invitrogen™

Qubit™ Flex Fluorometers and Starter Kits

Experience accurate, specific, and sensitive quantification of DNA, RNA, microRNA, protein, and endotoxin with the Qubit Flex Fluorometer. Measure up to eight samples simultaneously using Qubit assay kits for reproducible results in seconds.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeWarranty
A45924NGS Starter Kit3-year Rapid Exchange Warranty
A44984Benchtop Fluorometer3-year Rapid Exchange Warranty
A44985Benchtop Fluorometer3-year AB Repair Center Warranty
Q33327FluorometerStandard Warranty
A45925Quantitation Starter Kit3-year Rapid Exchange Warranty
A45927Quantitation Starter Kit3-year AB Repair Center Warranty
Q45894Quantitation Starter KitStandard Warranty
A45926NGS Starter Kit3-year AB Repair Center Warranty
Q45893NGS Starter KitStandard Warranty
Catalog number A45924
Price (EUR)
7.261,78
Each
Type:
NGS Starter Kit
Warranty:
3-year Rapid Exchange Warranty
Price (EUR)
7.261,78
Each

The Qubit Flex Fluorometer is designed for highly accurate and sensitive quantification of DNA, RNA, microRNA, protein, and endotoxin. This benchtop fluorometer offers the flexibility to directly measure the fluorescence of up to eight samples simultaneously, which reduces assay variability. Paired with Qubit reagents, the Qubit Flex Fluorometer is part of an optimized workflow generating highly accurate and reproducible results. The fluorometer has a large screen and intuitive user interface, making it easy to select and run your desired assay with results displayed in just a few seconds.

Features of the Qubit Flex Fluorometer include:

  • Improved throughput—measure multiple samples and quantify DNA, RNA, protein, and endotoxin up to 50% faster than single-sample readers
  • Flexibility—measure up to eight samples simultaneously using 1–20 μL of sample
  • Complete optimized system—works seamlessly with Qubit reagents to generate reliable, sensitive, and specific results
  • On-board calculators—quickly generates Qubit working solution preparation instructions and determines the molarity of your samples
  • Easy to use—a large 8-inch, state-of-the-art color touchscreen and intuitive user interface results in easy workflow navigation
  • Data export flexibility—data is transferred easily as a CSV or PDF using the included Wi-Fi dongle, a USB drive, or connection to your network drive
  • Several languages available—choices include English, French, Spanish, Italian, German, simplified Chinese, and Japanese
  • Verification test—a pre-loaded program is included to verify performance of the instrument
  • Qubit Flex SmartStart Digital Educational Experience—includes interactive visual demonstrations of how to install, operate, and maintain your instrument

Higher throughput, less variability than single sample readers

The Qubit Flex Fluorometer has the flexibility to measure multiple DNA, RNA, protein, and endotoxin samples up to 50% faster than single-sample readers. In addition to saving time, the ability to measure eight samples simultaneously reduces variability, resulting in highly reproducible data. The fluorometer includes reagent, range, and molarity calculators, further reducing the time to results. The reagent and range calculators help to determine which reagent and what amount of sample to use to effectively quantify the DNA, RNA, or protein. The molarity calculator allows you to calculate the molarity of your samples based on nucleic acid length and the measured concentration.

Enables more accurate results than UV absorbance at low concentrations

The Qubit Flex Fluorometer is far more sensitive than UV absorbance, which measures anything absorbing at 260 nm, including DNA, RNA, protein, free nucleotides, and excess salts. Moreover, UV spectrophotometry is often not sensitive enough to accurately measure low concentrations of DNA and RNA. With the Qubit Flex Fluorometer, your research is enhanced by more accurate measurements because the dyes in the Qubit assay kits fluoresce only when bound to the target molecule (DNA, RNA, protein, or endotoxin) in your sample. This allows you to avoid repeating work due to inaccurate measurements.

Comprehensive warranty options

The Qubit Flex Fluorometer comes with a standard 1-year warranty that covers repair costs including shipping, parts, and labor. Service plans can be bundled with new instrument purchases to provide a 3-year warranty. These Qubit Flex Fluorometer packages are offered with two different types of bundled warranties:

  • Rapid Exchange (REX) warranty (3-year)
    • Replacement shipped out in ~1 business day for labs with critical timelines
    • Available with the Qubit Flex Fluorometer (Cat. No. A44984), Qubit Flex NGS Starter Kit (Cat. No. A45924), Qubit Flex Quantification Starter Kit (Cat. No. A45925), Qubit Flex Endotoxin Starter Kit (Cat. No. Q32894REXPKG) and Qubit Flex Fluorometer with Security Software (Cat. No. Q45895REXPKG)
  • AB Repair Center (ABRC) warranty (3-year)
    • Instrument repaired and returned in ~3 weeks, ideal for regulated labs required to maintain asset tagging consistency
    • Available with the Qubit Flex Fluorometer (Cat. No. A44985), Qubit Flex NGS Starter Kit (Cat. No. A45926), Qubit Flex Quantification Starter Kit (Cat. No. A45927), Qubit Flex Endotoxin Starter Kit (Cat. No. Q32894PKG) and Qubit Flex Fluorometer with Security Software (Cat. No. Q45895PKG)

21 CFR Part 11 Software Module

The Qubit Flex Security Software supports compliance with 21 CFR Part 11 FDA guidelines for security, auditing, and electronic (SAE) signatures using the SAE administration console. The 21 CFR Part 11 software can be purchased separately (Cat. No. Q31994).

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Emission Wavelength513-563 nm (Green), 671-693 nm (Far-red)
IncludesQubit Flex NGS Starter Kit with 3-year Rapid Exchange Warranty
Warranty3-year Rapid Exchange Warranty
Excitation Wavelength460 to 480 nm (Blue LED), 620 to 640 nm (Red LED)
For Use With (Equipment)Qubit Flex Fluorometer
Product LineQubit
TypeNGS Starter Kit
Wavelength Range400 to 750 nm
Unit SizeEach
Package Components
Qubit™ Flex NGS Starter Kit
Catalog Number
Q45893
Unit Size
1 kit
Qty
1
SmartStart Qubit™ Flex Digital
Catalog Number
TRN00304
Unit Size
Each
Qty
1
Rapid Exchange, Qubit™ Flex
Catalog Number
ZGEXSCQFLEX1Y
Unit Size
Each
Qty
2
Contents & Storage
Qubit Flex Fluorometers include:
•Qubit Flex Fluorometer (Cat. No. Q33327)
•WiFi Dongle (Cat. No. A26774)
•Qubit Flex SmartStart Digital Educational Experience—includes interactive visual demonstrations of how to install, operate, and maintain your instrument

Qubit Flex Quantitation Starter Kits include:
•Qubit Flex Fluorometer (Cat. No. Q33327)
•Qubit Flex Assay Tube Strips, 125 strips (Cat. No. Q33252)
•Qubit 1X dsDNA HS Assay Kit, 100 assays (Cat. No. Q33230)
•Qubit 1X dsDNA HS Assay Kit, 100 assays (Cat. No. Q33265)
•Qubit RNA HS Assay Kit, 100 assays (Cat. No. Q32852)
•Qubit RNA BR Assay Kit, 100 assays (Cat. No. Q10210)
•Qubit Protein Assay Kit, 100 assays (Cat. No. Q33211)
•Qubit Flex System Verification Assay Kit (Cat. No. Q33254)
•Qubit Flex Reservoirs, 100 reservoirs (Cat. No. Q33253)
•WiFi Dongle (Cat. No. A26774)
•Qubit Flex SmartStart Digital Educational Experience—includes interactive visual demonstrations of how to install, operate, and maintain your instrument

Qubit Flex NGS (Next-Generation Sequencing) Starter Kit includes:
•Qubit Flex Fluorometer (Cat. No. Q33327)
•Qubit Flex Assay Tubes Strips, 125 strips (Cat. No. Q33252)
•Qubit 1X dsDNA HS Assay Kit, 500 assays (Cat. No. Q33231)
•Qubit Flex Assay Reservoirs, 100 reservoirs (Cat. No. Q33253)
•WiFi Dongle (Cat. No. A26774)
•Qubit Flex SmartStart Digital Educational Experience—includes interactive visual demonstrations of how to install, operate, and maintain your instrument