Library Index Adapter Kit
Applied Biosystems™

Library Index Adapter Kit

The Library Index Adapter Kit provides the indexing components required for amplified library preparation with the Enzymatic Fragmentation DNA Library Prep with Amp Mix.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMultiplex Capability
A5057706Set 3 of 96 Unique Indexes
A505770416 Unique Indexes
A5057708Set 1 of 96 Unique Indexes
A5057707Set 2 of 96 Unique Indexes
A5057705Set 4 of 96 Unique Indexes
Catalog number A5057706
Price (EUR)
430,00
Each
Multiplex Capability:
Set 3 of 96 Unique Indexes
Price (EUR)
430,00
Each

The Library Index Adapter Kit provides the indexing components required for amplified library preparation with the Enzymatic Fragmentation DNA Library Prep with Amp Mix. The kit supplies Universal Adapters for ligation and Dual Index Primers for PCR amplification, enabling generation of multiplex-ready libraries for next-generation sequencing (NGS) workflows.

Key features:

  • Base-balanced primer layout to support pooled next-generation sequencing workflows
  • Compatible with downstream hybridization-based target enrichment workflows
  • Dual Index Primers for sample-specific indexing during amplification
  • One reaction per index pair

Two-component indexing strategy. Universal Adapters are ligated after fragmentation, end-repair, and dA-tailing, then unique Dual Index Primers are added during PCR amplification to generate amplified, indexed libraries.

Designed for multiplexed workflows. The Dual Index Primers are supplied in a plate with one reaction per index pair and are base-balanced on a column basis. For 8-plex hybridization, libraries should be selected from a single column to maintain balanced pooling.

Workflow integration. This kit is used with the Enzymatic Fragmentation DNA Library Prep with Amp Mix and feeds directly into standard or quick target enrichment workflows. Universal Adapters are packaged separately within the box for convenient handling.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Barcode10 basepair index length
Certifications/ComplianceISO 9001, ISO 13485
For Use With (Equipment)Short Read NGS
IncludesDual Index Primers, Universal Adapters
Multiplex CapabilitySet 3 of 96 Unique Indexes
No. of Samples96 Samples
Nucleic Acid TypeDNA
OverhangT-A
Plate Type96-well Plate
Product TypeLibrary Adapter
Regulatory StatusRUO
Sample TypeGenomic DNA
Shelf Life12 Months
Workflow StepLibrary Preparation
FormatKit
Isolation TechnologyMagnetic Bead
Sequencing TypeShort Read Sequencing
Unit SizeEach
Contents & Storage
-20°C