Contents & Storage
• Test plates, 5 x 12-strip plates
• Positive control (PC), 600 μL
• Negative control (NC), 600 μL
• ASFV sample dilution buffer, 150 mL
• ASFV conjugate, 60 mL
• Chromogen (TMB) substrate, 60 mL
• Stop solution, 60 mL
• Washing fluid (20X), 2 x 60 mL
• Plate sealers, 10
Store at 2–8°C.