2,5-Dimethyl-1,5-hexadien-3-ol, 97%
2,5-Dimethyl-1,5-hexadien-3-ol, 97%
2,5-Dimethyl-1,5-hexadien-3-ol, 97%
Thermo Scientific Chemicals

2,5-Dimethyl-1,5-hexadien-3-ol, 97%

CAS: 17123-63-6 | C8H14O | 126.199 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B20072.14
also known as B20072-14
Price (EUR)
157,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
157,00
Each
Chemical Identifiers
CAS17123-63-6
IUPAC Name(3S)-2,5-dimethylhexa-1,5-dien-3-ol
Molecular FormulaC8H14O
InChI KeyQQMFDURNWCUKNS-QMMMGPOBSA-N
SMILESCC(=C)C[C@H](O)C(C)=C
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4555-1.4605 @ 20?C
FormLiquid
Appearance (Color)Clear colorless
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only