You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
Home
▸
Shop All Products
▸
Chemicals
▸
Organic compounds
▸
Hydrocarbons
▸
Saturated hydrocarbons
▸
2,2,3-Trimethylbutane, 98%
Element Search
Structure Search
Thermo Scientific™
2,2,3-Trimethylbutane, 98%
CAS: 464-06-2 | C7H16 | 100.21 g/mol
Have Questions?
Change view
Quantity:
1 g
5 g
25 g
Try the chem dex
TM
web app
Catalog number
B20205.03
also known as B20205-03
Price (EUR)
22,40
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (EUR)
22,40
Each
Add to cart
Chemical Identifiers
CAS
464-06-2
IUPAC Name
2,2,3-trimethylbutane
Molecular Formula
C7H16
InChI Key
ZISSAWUMDACLOM-UHFFFAOYSA-N
SMILES
CC(C)C(C)(C)C
View more
Specifications
Specification Sheet
Appearance (Color)
Clear colorless
Assay (GC)
≥97.5%
Form
Liquid
Refractive Index
1.3880-1.3900 @ 20?C