3,5-Bis(trifluoromethyl)phenyl isothiocyanate, 98%
3,5-Bis(trifluoromethyl)phenyl isothiocyanate, 98%
3,5-Bis(trifluoromethyl)phenyl isothiocyanate, 98%
3,5-Bis(trifluoromethyl)phenyl isothiocyanate, 98%
Thermo Scientific Chemicals

3,5-Bis(trifluoromethyl)phenyl isothiocyanate, 98%

CAS: 23165-29-9 | C9H3F6NS | 271.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B20206.06
also known as B20206-06
Price (EUR)
93,50
Online Exclusive
110,00
Save 16,50 (15%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (EUR)
93,50
Online Exclusive
110,00
Save 16,50 (15%)
Each
Add to cart
Chemical Identifiers
CAS23165-29-9
IUPAC Name1-isothiocyanato-3,5-bis(trifluoromethyl)benzene
Molecular FormulaC9H3F6NS
InChI KeyFXOSSGVJGGNASE-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC(=CC(=C1)N=C=S)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
FormLiquid
Refractive Index1.4975-1.5015 @ 20?C
Appearance (Color)Clear colorless to yellow
Assay (GC)≥97.5%
It is used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an active pharmaceutical intermediate.

Solubility
Hydrolyzes in water.

Notes
Moisture sensitive. Store away from alcohols, amines and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from water/moisture.
RUO – Research Use Only

General References:

  1. Marcus Frings, et al. Synthesis of chiral sulfoximine-based thioureas and their application in asymmetric organocatalysis.Beilstein J Org Chem.2012,81443-1451.
  2. Paul M Dietrich, et al. Self-assembled monolayers of aromatic μ-aminothiols on gold: surface chemistry and reactivity.Langmuir.2010,26(6), 3949-3954.