1-Methylcycloheptanol, 98%, Thermo Scientific Chemicals
1-Methylcycloheptanol, 98%, Thermo Scientific Chemicals
1-Methylcycloheptanol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Methylcycloheptanol, 98%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
B20393.06
also known as B20393-06
5 g
Catalog number B20393.06
also known as B20393-06
Price (EUR)
146,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
146,00
Each
Chemical Identifiers
CAS3761-94-2
IUPAC Name1-methylcycloheptan-1-ol
Molecular FormulaC8H16O
InChI KeyXFFKAYOHINCUNU-UHFFFAOYSA-N
SMILESCC1(O)CCCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Refractive Index1.4700-1.4740 @ 20?C
FormLiquid
Assay (GC)≥97.5%
1-Methylcycloheptanol, is used as an pharmaceutical and chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methylcycloheptanol, is used as an pharmaceutical and chemical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

General References:

  1. R Kotani. Demjanov Rearrangement of 1-Methylcyclohexanemethylamine. The Journal of Organic Chemistry.196530 (2), 350-354.
  2. HC Brown.; M Borkowski. The Effect of Ring Size on the Rate of Solvolysis of the 1-Chloro-1-methylcycloalkanes1, 2. J. Am. Chem. Soc. 195274 (8), 1894-1902.