5-Chlorothiophene-2-sulfonamide, 97%
5-Chlorothiophene-2-sulfonamide, 97%
5-Chlorothiophene-2-sulfonamide, 97%
Thermo Scientific Chemicals

5-Chlorothiophene-2-sulfonamide, 97%

CAS: 53595-66-7 | C4H4ClNO2S2 | 197.65 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B20477.03
also known as B20477-03
Price (EUR)
14,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
14,00
Each
Chemical Identifiers
CAS53595-66-7
IUPAC Name5-chlorothiophene-2-sulfonamide
Molecular FormulaC4H4ClNO2S2
InChI KeyRKLQLYBJAZBSEU-UHFFFAOYSA-N
SMILESNS(=O)(=O)C1=CC=C(Cl)S1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)110.0-120.0?C
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only