2,6-Dichloro-3-nitrotoluene, 99%
2,6-Dichloro-3-nitrotoluene, 99%
2,6-Dichloro-3-nitrotoluene, 99%
Thermo Scientific Chemicals

2,6-Dichloro-3-nitrotoluene, 99%

CAS: 29682-46-0 | C7H5Cl2NO2 | 206.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number B20520.09
also known as B20520-09
Price (EUR)
30,60
Each
Quantity:
10 g
Request bulk or custom format
Price (EUR)
30,60
Each
Chemical Identifiers
CAS29682-46-0
IUPAC Name1,3-dichloro-2-methyl-4-nitrobenzene
Molecular FormulaC7H5Cl2NO2
InChI KeyWBNZUUIFTPNYRN-UHFFFAOYSA-N
SMILESCC1=C(Cl)C=CC(=C1Cl)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.5%
FormPowder or lumps
Appearance (Color)Pale yellow
Melting Point (clear melt)51.0-58.0?C
2,6-Dichloro-3-nitrotoluene, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dichloro-3-nitrotoluene, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. EE Haley.; JP Lambooy. The Synthesis of 6-Chloro-7-methyl-9-(1'-D-ribityl)-isoalloxazine and 6-Methyl-7-chloro-9-(1'-D-ribityl)-isoalloxazine1. J. Am. Chem. Soc. 195476 (20), 5093-5096.
  2. G Pozzi.; I Colombani.; M Miglioli.; F Montanari.; S Quici. Epoxidation of Alkenes Under Liquid-Liquid Biphasic Conditions: Synthesis and Catalytic Activity of Mn (III)-Tetraarylporphyrins Bearing Perfluoroalkyl Tails. Tetrahedron. 199753 (17), 6145-6162.