Thermo Scientific Chemicals

4-Ethylbenzaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: B20645.14
25 g, Each
Thermo Scientific Chemicals

4-Ethylbenzaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: B20645.14
25 g, Each
Quantity
Catalog number: B20645.14
also known as B20645-14
Price (EUR)
Price: 39,00
Online price: 35,10
Your price:
Quantity
-

Chemical Identifiers

CAS
4748-78-1
IUPAC Name
4-ethylbenzaldehyde
Molecular Formula
C9H10O
InChI Key
QNGNSVIICDLXHT-UHFFFAOYSA-N
SMILES
CCC1=CC=C(C=O)C=C1
Form
Liquid
Refractive Index
1.5365-1.5415 @ 20?C
Assay (GC)
≥96.0%
Appearance (Color)
Clear colorless to pale yellow
Free acid (titration)
≤3.0%

Description

It is used as a perfuming agent in cosmetics industry. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a perfuming agent in cosmetics industry. It is also used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix in water. Soluble in alcohol.

Notes
Air sensitive. Store away from oxidizing agents and air. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 4
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H302 - Harmful if swallowed

    EU Specific Hazard Statements

    Combustible liquid

    Precautionary Statements

    P264 - Wash face, hands and any exposed skin thoroughly after handling

    P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

    P312 - Call a POISON CENTER or doctor if you feel unwell