n-Propyl propionate, 99%
n-Propyl propionate, 99%
n-Propyl propionate, 99%
n-Propyl propionate, 99%
Thermo Scientific Chemicals

n-Propyl propionate, 99%

CAS: 106-36-5 | C6H12O2 | 116.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1000 mL
Catalog number B20680.AU
also known as B20680-AU
Price (EUR)
130,90
Online Exclusive
154,00
Save 23,10 (15%)
Each
Add to cart
Quantity:
1000 mL
Request bulk or custom format
Price (EUR)
130,90
Online Exclusive
154,00
Save 23,10 (15%)
Each
Add to cart
Chemical Identifiers
CAS106-36-5
IUPAC Namepropyl propanoate
Molecular FormulaC6H12O2
InChI KeyMCSINKKTEDDPNK-UHFFFAOYSA-N
SMILESCCCOC(=O)CC
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless
Assay (GC)≥98.5% (UK-sourced material)
Refractive Index1.3915-1.3950 @ 20?C (UK-sourced material)
Assay from Supplier's CofA≥98.5% (US-sourced material)
View more
It has application in automotive refinish, OEM coatings, appliance coatings, printing inks and as polymerization solvent. Its good activity yields efficient resin viscosity reduction that enables the development of lower VOC coatings. Its high electrical resistivity is beneficial in optimizing transfer efficiency in coatings applied via electrostatic spray equipment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It has application in automotive refinish, OEM coatings, appliance coatings, printing inks and as polymerization solvent. Its good activity yields efficient resin viscosity reduction that enables the development of lower VOC coatings. Its high electrical resistivity is beneficial in optimizing transfer efficiency in coatings applied via electrostatic spray equipment.

Solubility
Soluble in water. (5g/L).

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Ernesto Altman.;Georgios D. Stefanidis.; Thomas van Gerven.; Andrzej I. Stankiewicz. Process Intensification of Reactive Distillation for the Synthesis of n-Propyl Propionate: The Effects of Microwave Radiation on Molecular Separation and Esterification Reaction. Ind. Eng. Chem. Res. 2010, 49 (21),10287-10296 .
  2. Toshio Hayashi.; Takahiro Inagaki.; Naohiko Itayama.; Hideyuki Baba. Selective oxidation of alcohol over supported gold catalysts: methyl glycolate formation from ethylene glycol and methanol. Catal. Today. 2006, 117 (1-3),210-213 .