Cyclopropyl 4-methoxyphenyl ketone, 98%
Cyclopropyl 4-methoxyphenyl ketone, 98%
Cyclopropyl 4-methoxyphenyl ketone, 98%
Thermo Scientific Chemicals

Cyclopropyl 4-methoxyphenyl ketone, 98%

CAS: 7152-03-6 | C11H12O2 | 176.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B20725.06
also known as B20725-06
Price (EUR)
28,40
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
28,40
Each
Specifications
Beilstein2046539
CAS7152-03-6
Chemical Name or MaterialCyclopropyl 4-methoxyphenyl ketone
EINECS Number230-487-7
Flash Point>110°C (230°F)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only