4,4'-Diisopropylbiphenyl, 97%
4,4'-Diisopropylbiphenyl, 97%
4,4'-Diisopropylbiphenyl, 97%
Thermo Scientific Chemicals

4,4'-Diisopropylbiphenyl, 97%

CAS: 18970-30-4 | C18H22 | 238.37 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B20779.03
also known as B20779-03
Price (EUR)
32,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
32,00
Each
Chemical Identifiers
CAS18970-30-4
IUPAC Name4,4'-bis(propan-2-yl)-1,1'-biphenyl
Molecular FormulaC18H22
InChI KeyNUEUMFZLNOCRCQ-UHFFFAOYSA-N
SMILESCC(C)C1=CC=C(C=C1)C1=CC=C(C=C1)C(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream or pale yellow
FormPowder or crystalline powder or crystals
Assay (GC)≥96.0%
Melting Point62-68°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only