1-Iodopentane (pentyl iodide) was used in the preparation of 1-pentyl radical by shock heating process.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Iodopentane (pentyl iodide) was used in the preparation of 1-pentyl radical by shock heating process.
Solubility
Insoluble in water.
Notes
Light sensitive. Store in dark place. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
- F. Grane. Measurements of Transverse Dispersion in Granular Media. J. Chem. Eng. Data. 1961, 6 (2), 283-287 .
- Margaret Shea Chiles.; Donald D. Jackson.; Perry C. Reeves. Preparation and synthetic utility of phase-transfer catalysts anchored to polystyrene.J. Org. Chem. 1980, 45 (14),2915-2918 .