1-(4-Methylphenyl)ethanol, 97%
1-(4-Methylphenyl)ethanol, 97%
1-(4-Methylphenyl)ethanol, 97%
1-(4-Methylphenyl)ethanol, 97%
Thermo Scientific Chemicals

1-(4-Methylphenyl)ethanol, 97%

CAS: 536-50-5 | C9H12O | 136.194 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B20832.06
also known as B20832-06
Price (EUR)
95,80
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (EUR)
95,80
Each
Add to cart
Chemical Identifiers
CAS536-50-5
IUPAC Name1-(4-methylphenyl)ethan-1-ol
Molecular FormulaC9H12O
InChI KeyJESIHYIJKKUWIS-UHFFFAOYNA-N
SMILESCC(O)C1=CC=C(C)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.5190-1.5240 @ 20?C
Appearance (Color)Clear colorless
1-(4-Methylphenyl)ethanol serves as a component of the essential oil from some plants in the ginger family. It is also used as a flavoring agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(4-Methylphenyl)ethanol serves as a component of the essential oil from some plants in the ginger family. It is also used as a flavoring agent.

Solubility
Slightly miscible with water.

Notes
Incompatible with oxidizing agents and strong reducing agents. Store in a cool place.
RUO – Research Use Only

General References:

  1. Reddy, M. M.; Swamy, P.; Naresh, M.; Srujana, K.; Durgaiah, C.; Rao, T. V.; Narender, N. One-pot synthesis of alpha-iodoketones from alcohols using ammonium iodide and Oxone in water. RSC Adv. 2015, 5 (16), 12186-12190.
  2. Zhang, Y.; Zhou, Q.; Ma, W.; Zhao, J. Enantioselective oxidation of racemic secondary alcohols catalyzed by chiral Mn(III)-salen complex with sodium hypochlorite as oxidant. Catal. Commun. 2014, 45, 114-117.