1,2,3-Thiadiazole-4-carboxylic acid, 97%
1,2,3-Thiadiazole-4-carboxylic acid, 97%
1,2,3-Thiadiazole-4-carboxylic acid, 97%
Thermo Scientific Chemicals

1,2,3-Thiadiazole-4-carboxylic acid, 97%

CAS: 4100-13-4 | C3H2N2O2S | 130.121 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B20842.03
also known as B20842-03
Price (EUR)
87,60
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
87,60
Each
Chemical Identifiers
CAS4100-13-4
IUPAC Name1,2,3-thiadiazole-4-carboxylate
Molecular FormulaC3HN2O2S
InChI KeyHJZYBDPHAHGHAZ-UHFFFAOYSA-M
SMILES[O-]C(=O)C1=CSN=N1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (HPLC)≥96.0%
Assay (Aqueous acid-base Titration)≥96.0% to ≤104.0%
Appearance (Color)Pale brown to brown
It is used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an active pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents, reducing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Quan, Mimi L.; Hu, Zilun; Wang, Cailan. Tricyclic pyrido-​carboxamide derivatives as rock inhibitors and their preparation.PCT Int. Appl.2015.
  2. Zaifeng Li, et al. Synthesis and Antifungal Activities of Alkyl N-(1,2,3-Thiadiazole-4-Carbonyl) Carbamates and S-Alkyl N-(1,2,3-Thiadiazole-4-Carbonyl) Carbamothioates.. Agric. Food Chem.,2005,53(10), 3872-3876.