5-(2-Pyridyl)-1H-tetrazole, 98%
5-(2-Pyridyl)-1H-tetrazole, 98%
5-(2-Pyridyl)-1H-tetrazole, 98%
Thermo Scientific Chemicals

5-(2-Pyridyl)-1H-tetrazole, 98%

CAS: 33893-89-9 | C6H4N5 | 146.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B20852.03
also known as B20852-03
Price (EUR)
106,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
106,00
Each
Chemical Identifiers
CAS33893-89-9
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Appearance (Color)White to pale cream or pale pink
Identification (FTIR)Conforms
Assay (HPLC)≥97.5%
It is used in the synthesis and crystal structure of the [Ag(2-allyl-5-phenyl-2Htetrazole)ClO4] π,complex .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the synthesis and crystal structure of the [Ag(2-allyl-5-phenyl-2Htetrazole)ClO4] π,σ-complex .

Solubility
Soluble in water at 25°C 11 g/L.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Xiao-Jie Mo, et al. An unexpected in situ formation of a substitute tetrazole ligand and its Mn(II) and Cu(II) complexes.Inorg. Chem. Commun.,2004,7(3), 353-355.
  2. Dr. Adina Morozan, et al. Metal-Free Nitrogen-Containing Carbon Nanotubes Prepared from Triazole and Tetrazole Derivatives Show High Electrocatalytic Activity towards the Oxygen Reduction Reaction in Alkaline Media.ChemSusChem.,2012,5(4), 647-651.