3-Methylbenzyl alcohol, 98%, Thermo Scientific Chemicals
3-Methylbenzyl alcohol, 98%, Thermo Scientific Chemicals
3-Methylbenzyl alcohol, 98%, Thermo Scientific Chemicals
3-Methylbenzyl alcohol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Methylbenzyl alcohol, 98%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
B20875.1425 g
Catalog number B20875.14
Price (EUR)
54,49
Online Exclusive
64,10
Save 9,61 (15%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
54,49
Online Exclusive
64,10
Save 9,61 (15%)
Each
Add to cart
Chemical Identifiers
CAS587-03-1
IUPAC Name(3-methylphenyl)methanol
Molecular FormulaC8H10O
InChI KeyJJCKHVUTVOPLBV-UHFFFAOYSA-N
SMILESCC1=CC=CC(CO)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.0%
Appearance (Color)Clear colorless
FormLiquid
Refractive Index1.5315-1.5365 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only