n-Propyltriethoxysilane, 97%
n-Propyltriethoxysilane, 97%
n-Propyltriethoxysilane, 97%
Thermo Scientific Chemicals

n-Propyltriethoxysilane, 97%

CAS: 2550-02-9 | C9H22O3Si | 206.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number B20997.22
also known as B20997-22
Price (EUR)
99,65
온라인 행사
110,00
Save 10,35 (9%)
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
99,65
온라인 행사
110,00
Save 10,35 (9%)
Each
Chemical Identifiers
CAS2550-02-9
IUPAC Nametriethoxy(propyl)silane
Molecular FormulaC9H22O3Si
InChI KeyNBXZNTLFQLUFES-UHFFFAOYSA-N
SMILESCCC[Si](OCC)(OCC)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥96.0% (UK-sourced material)
FormLiquid
Assay from Supplier's CofA≥96.0% (US-sourced material)
Refractive Index1.3945-1.3995 @ 20?C (UK-sourced material)
View more
It is an important material in sol-gel production, it provides a hydrophobic surface treatment for inorganic powders or filter materials and it is one of the catalyst components for polyolefin production with Ziegler-Natta catalyst. It is mainly used as construction waterproofing agent/protective agent, inorganic filler surface treatment agent, and pigment dispersant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important material in sol-gel production, it provides a hydrophobic surface treatment for inorganic powders or filter materials and it is one of the catalyst components for polyolefin production with Ziegler-Natta catalyst. It is mainly used as construction waterproofing agent/protective agent, inorganic filler surface treatment agent, and pigment dispersant.

Solubility
Not miscible or difficult to mix in water. Soluble in alcohol, aromatic and aliphatic hydrocarbons.

Notes
Moisture sensitive. Store away from air and oxidizing agents and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep ignition sources away.
RUO – Research Use Only

General References:

  1. Emi Ukaji, et al. The effect of surface modification with silane coupling agent on suppressing the photo-catalytic activity of fine TiO2 particles as inorganic UV filter.Appl. Surf. Sci.,2007254(2), 563-569.
  2. G. Falini, et al. Protein crystallisation on chemically modified mica surfaces.Acta Cryst.,2002,D581649-1652.