Ethyl 7-methoxybenzo[b]furan-2-carboxylate, 97%
Ethyl 7-methoxybenzo[b]furan-2-carboxylate, 97%
Ethyl 7-methoxybenzo[b]furan-2-carboxylate, 97%
Thermo Scientific Chemicals

Ethyl 7-methoxybenzo[b]furan-2-carboxylate, 97%

CAS: 50551-58-1 | C12H12O4 | 220.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B21066.03
also known as B21066-03
Price (EUR)
22,60
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
22,60
Each
Chemical Identifiers
CAS50551-58-1
IUPAC Nameethyl 7-methoxy-1-benzofuran-2-carboxylate
Molecular FormulaC12H12O4
InChI KeySXCMZYVXJUJIDJ-UHFFFAOYSA-N
SMILESCCOC(=O)C1=CC2=CC=CC(OC)=C2O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale yellow
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Melting Point (clear melt)83.0-93.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only