2-Methoxybenzeneboronic acid, 97%
2-Methoxybenzeneboronic acid, 97%
2-Methoxybenzeneboronic acid, 97%
Thermo Scientific Chemicals

2-Methoxybenzeneboronic acid, 97%

CAS: 5720-06-9 | C7H9BO3 | 151.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B21071.14
also known as B21071-14
Price (EUR)
288,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
288,00
Each
Chemical Identifiers
CAS5720-06-9
IUPAC Name(2-methoxyphenyl)boronic acid
Molecular FormulaC7H9BO3
InChI KeyROEQGIFOWRQYHD-UHFFFAOYSA-N
SMILESCOC1=CC=CC=C1B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥96.0%
Proton NMRConforms to structure
Assay (HPLC)≥96.0%
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
2-Methoxylphenylboronic acid is a phenylboronic acid used to investigate boron function in plants. It is used in Suzuki reaction, organic reaction that is classified as a coupling reaction where the coupling partners are a boronic acid with a halide catalyzed by a palladium(0) complex.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methoxylphenylboronic acid is a phenylboronic acid used to investigate boron function in plants. It is used in Suzuki reaction, organic reaction that is classified as a coupling reaction where the coupling partners are a boronic acid with a halide catalyzed by a palladium(0) complex.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Jun Yan, et al. The relationship among pKa, pH, and binding constants in the interactions between boronic acids and diols—it is not as simple as it appears.Tetrahedron.200460(49), 11205-11209.
  2. Kazuaki Ishihara, et al. Bronsted Acid Assisted Chiral Lewis Acid (BLA) Catalyst for Asymmetric Diels-Alder Reaction.J. Am. Chem. Soc.,1994116(4), 1561-1562.