(±)-2-Pentanol, 99%
(±)-2-Pentanol, 99%
(±)-2-Pentanol, 99%
Thermo Scientific Chemicals

(±)-2-Pentanol, 99%

CAS: 6032-29-7 | C5H12O | 88.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2500 mL
Catalog number B21217.0F
also known as B21217-0F
Price (EUR)
218,65
キャンペーン価格
243,00
Save 24,35 (10%)
Each
Quantity:
2500 mL
Request bulk or custom format
Price (EUR)
218,65
キャンペーン価格
243,00
Save 24,35 (10%)
Each
Chemical Identifiers
CAS6032-29-7
IUPAC Namepentan-2-ol
Molecular FormulaC5H12O
InChI KeyJYVLIDXNZAXMDK-UHFFFAOYNA-N
SMILESCCCC(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥98.5%
Identification (FTIR)Conforms
Refractive Index1.4050-1.4080 @ 20?C
FormLiquid
It finds its application as a solvent for paints and lacquers and to make drugs, to make catalyst systems and methanol and as a solvent for primers and for extracting 2-hydroxy-4-methylthiobutyric acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application as a solvent for paints and lacquers and to make drugs, to make catalyst systems and methanol and as a solvent for primers and for extracting 2-hydroxy-4-methylthiobutyric acid.

Solubility
Soluble in water at 20°C, 166g/L. Soluble in alcohol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. G Silva-Oliver, et al. Vapor-liquid equilibria and critical points for the carbon dioxide +1-pentanol and carbon dioxide +2-pentanol systems at temperatures from 332 to 432 K.Fluid Phase Equilib.2002200(1), 161-172.
  2. Shaheen A. Al-Muhtaseb, et al. Phase equilibria of the ternary system water/acetic acid/2-pentanol.Fluid Phase Equilib.1996123(1-2), 189-203.