alpha-D-(+)-Talose, 97%
alpha-D-(+)-Talose, 97%
alpha-D-(+)-Talose, 97%
Thermo Scientific Chemicals

alpha-D-(+)-Talose, 97%

CAS: 2595-98-4 | C6H12O6 | 180.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.1 g
0.5 g
Catalog number B21226.77
also known as B21226-77
Price (EUR)
99,65
Online exclusive
111,00
Save 11,35 (10%)
Each
Quantity:
0.1 g
Request bulk or custom format
Price (EUR)
99,65
Online exclusive
111,00
Save 11,35 (10%)
Each
Chemical Identifiers
CAS2595-98-4
IUPAC Name2,3,4,5,6-pentahydroxyhexanal
Molecular FormulaC6H12O6
InChI KeyGZCGUPFRVQAUEE-UHFFFAOYNA-N
SMILESOCC(O)C(O)C(O)C(O)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Melting Point (clear melt)125.0-131.0?C
Optical Rotation21? ± 1.0? (c=1 in water)
Assay (Silylated GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only