1,4-Dichlorobutane, 97%
1,4-Dichlorobutane, 97%
1,4-Dichlorobutane, 97%
Thermo Scientific Chemicals

1,4-Dichlorobutane, 97%

CAS: 110-56-5 | C4H8Cl2 | 127.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number B21337.30
also known as B21337-30
Price (EUR)
41,70
Each
Quantity:
250 g
Request bulk or custom format
Price (EUR)
41,70
Each
Chemical Identifiers
CAS110-56-5
IUPAC Name1,4-dichlorobutane
Molecular FormulaC4H8Cl2
InChI KeyKJDRSWPQXHESDQ-UHFFFAOYSA-N
SMILESClCCCCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.4520-1.4570 @ 20?C
Used in organic synthesis including adiponitrile. It is also used as an active pharmaceutical intermediate. 1,4-dichlorobutane (1,4-DCB) can be used as an organic solvent (O) for ion-transfer voltammetry at water (W) interface.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in organic synthesis including adiponitrile. It is also used as an active pharmaceutical intermediate. 1,4-dichlorobutane (1,4-DCB) can be used as an organic solvent (O) for ion-transfer voltammetry at water (W) interface.

Solubility
Soluble in water at 20°C2.4g/L.

Notes
Store away from oxidizing agents, alkali metals and alkali earth metals. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Hajime Katano, et al. Ion-transfer voltammetry at 1,6-dichlorohexane∣water and 1,4-dichlorobutane∣water interfaces.Talanta,200463(1), 185-193.
  2. Kirsten Aarset, et al. Molecular Structure and Conformational Composition of 1,4-Dichlorobutane and 1,4-Dibromobutane: a Gas-Phase Electron Diffraction and ab Initio Investigation.J. Phys. Chem. 199498(20), 5249-5256.