2,6-Dichlorophenylacetic acid, 98%
2,6-Dichlorophenylacetic acid, 98%
2,6-Dichlorophenylacetic acid, 98%
2,6-Dichlorophenylacetic acid, 98%
Thermo Scientific Chemicals

2,6-Dichlorophenylacetic acid, 98%

CAS: 6575-24-2 | C8H6Cl2O2 | 205.034 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B21354.14
also known as B21354-14
Price (EUR)
76,90
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
76,90
Each
Add to cart
Chemical Identifiers
CAS6575-24-2
IUPAC Name2-(2,6-dichlorophenyl)acetate
Molecular FormulaC8H5Cl2O2
InChI KeySFAILOOQFZNOAU-UHFFFAOYSA-M
SMILES[O-]C(=O)CC1=C(Cl)C=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (Silylated GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only