Cresol Red sodium salt
Cresol Red sodium salt
Cresol Red sodium salt
Thermo Scientific™

Cresol Red sodium salt

Transition interval (acid): pH 0.2 (red) to pH 1.8 (yellow); (alkaline) pH 7.2 (yellow) to pH 8.8 (red) | CAS: 62625-29-0 | C21H17NaO5S
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number B21361.14
also known as B21361-14
Price (EUR)
63,90
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
63,90
Each
Chemical Identifiers
CAS62625-29-0
IUPAC Namesodium 4-[3-(4-hydroxy-3-methylphenyl)-1,1-dioxo-3H-2,1λ⁶-benzoxathiol-3-yl]-2-methylbenzen-1-olate
Molecular FormulaC21H17NaO5S
InChI KeyFMKJVYROEYMYFV-UHFFFAOYNA-M
SMILES[Na+].CC1=CC(=CC=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C([O-])C(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange to red to dark red to brown to dark brown
FormPowder or crystalline powder and/or crystals
Color ChangeOrange @ pH <2.0 to yellow @ pH >3.0
Color ChangeYellow @ pH <7.2 to red/purple @ pH >8.8
Loss on Drying≤5.0% (110°C/1 hour)
View more
Cresol Red sodium salt acts as a constituent of polymerase chain reaction reagent systems. It is useful as a pH indicator with color changes with pH 0.2 - pH 1.8 and pH 7.1 - 8.8. Further, it plays an important role in molecular biology reactions in place of other loading dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cresol Red sodium salt acts as a constituent of polymerase chain reaction reagent systems. It is useful as a pH indicator with color changes with pH 0.2 - pH 1.8 and pH 7.1 - 8.8. Further, it plays an important role in molecular biology reactions in place of other loading dyes.

Solubility
Soluble in water, dilute alcohols, acids, alkalies and ethanol.

Notes
Transition interval (acid): pH 0.2 (red) to pH 1.8 (yellow); (alkaline) pH 7.2 (yellow) to pH 8.8 (red)Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Heck, M. V.; Azizov, M.; Stehning, T.; Walter, M.; Kedersha, N.; Auburger, G. Dysregulated expression of lipid storage and membrane dynamics factors in Tia1 knockout mouse nervous tissue. Neurogenetics 2014, 15 (2), 135-144.
  2. Mukherjee, S.; Hill, M. R.; Sumerlin, B. S. Self-healing hydrogels containing reversible oxime crosslinks. Soft matter 2015, 11 (30), 6152-6161.