cis-4-Cyclohexene-1,2-dicarboxylic anhydride, 95%
cis-4-Cyclohexene-1,2-dicarboxylic anhydride, 95%
cis-4-Cyclohexene-1,2-dicarboxylic anhydride, 95%
Thermo Scientific Chemicals

cis-4-Cyclohexene-1,2-dicarboxylic anhydride, 95%

CAS: 935-79-5 | C8H8O3 | 152.149 g/mol
Have Questions?
Catalog NumberQuantity
B21431.30
also known as B21431-30
250 g
Catalog number B21431.30
also known as B21431-30
Price (EUR)
27,60
Each
Quantity:
250 g
Request bulk or custom format
Price (EUR)
27,60
Each
Chemical Identifiers
CAS935-79-5
IUPAC Name(3aR,7aS)-1,3,3a,4,7,7a-hexahydro-2-benzofuran-1,3-dione
Molecular FormulaC8H8O3
InChI KeyKMOUUZVZFBCRAM-OLQVQODUSA-N
SMILESO=C1OC(=O)[C@@H]2CC=CC[C@H]12
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)97-104?C
Appearance (Color)White
FormFlakes
Identification (FTIR)Conforms
Assay (GC)≥94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only