Diethyl oxalacetate sodium salt, 95%
Diethyl oxalacetate sodium salt, 95%
Diethyl oxalacetate sodium salt, 95%
Thermo Scientific Chemicals

Diethyl oxalacetate sodium salt, 95%

CAS: 40876-98-0 | C8H11NaO5 | 210.161 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number B21486.36
also known as B21486-36
Price (EUR)
139,00
Each
Quantity:
500 g
Request bulk or custom format
Price (EUR)
139,00
Each
Chemical Identifiers
CAS40876-98-0
IUPAC Namesodium 1,4-diethoxy-1,3,4-trioxobutan-2-ide
Molecular FormulaC8H11NaO5
InChI KeyJPTKZRPOIUYFTM-UHFFFAOYSA-N
SMILES[Na+].CCOC(=O)[CH-]C(=O)C(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)Yellow or pale orange to brown or cream
Assay (Non-aqueous acid-base Titration)≥94.0%
Diethyl oxalacetate sodium salt is used in the preparation of oxaloacetic acid. It is also used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl oxalacetate sodium salt is used in the preparation of oxaloacetic acid. It is also used as an active pharmaceutical intermediate.

Solubility
Slightly soluble in water (1.2 g/L) (25°C).

Notes
Moisture sensitive. Store away from oxidizing agents and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
RUO – Research Use Only

General References:

  1. M F Utter, et al. Pyruvate Carboxylase. I. Nature Of The Reaction.J. Biol. Chem.19632382603-2608.
  2. Charles M. Hall, et al. Quinoline derivatives as antiallergy agents.J. Med. Chem., 197417(7), 685-690.