Methyl 4-fluorobenzoate, 97%
Methyl 4-fluorobenzoate, 97%
Methyl 4-fluorobenzoate, 97%
Thermo Scientific Chemicals

Methyl 4-fluorobenzoate, 97%

CAS: 403-33-8 | C8H7FO2 | 154.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B21504.06
also known as B21504-06
Price (EUR)
38,20
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
38,20
Each
Chemical Identifiers
CAS403-33-8
IUPAC Namemethyl 4-fluorobenzoate
Molecular FormulaC8H7FO2
InChI KeyMSEBQGULDWDIRW-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=C(F)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Refractive Index1.4925-1.4975 @ 20?C
Appearance (Color)Clear colorless
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only