Calix[6]arene, 98%
Calix[6]arene, 98%
Calix[6]arene, 98%
Thermo Scientific Chemicals

Calix[6]arene, 98%

CAS: 96107-95-8 | C42H36O6 | 636.744 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B21608.03
also known as B21608-03
Price (EUR)
59,65
Online exclusive
65,60
Save 5,95 (9%)
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
59,65
Online exclusive
65,60
Save 5,95 (9%)
Each
Chemical Identifiers
CAS96107-95-8
IUPAC Nameheptacyclo[31.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹.1²¹,²⁵.1²⁷,³¹]dotetraconta-1(37),3,5,7(42),9,11,13(41),15,17,19(40),21,23,25(39),27,29,31(38),33,35-octadecaene-37,38,39,40,41,42-hexol
Molecular FormulaC42H36O6
InChI KeyJLSWUKWFQCVKCL-UHFFFAOYSA-N
SMILESOC1=C2CC3=CC=CC(CC4=CC=CC(CC5=CC=CC(CC6=CC=CC(CC7=CC=CC(CC1=CC=C2)=C7O)=C6O)=C5O)=C4O)=C3O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Assay (HPLC)≥97.5%
Melting Point (DSC)360-385°C
FormPowder
It is used as a starting material for preparing functionalized calixarenes. It finds its uses as a pharmaceutical intermediate. It is also used in complexation and organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a starting material for preparing functionalized calixarenes. It finds its uses as a pharmaceutical intermediate. It is also used in complexation and organic synthesis.

Solubility
0.180g/100g of water @ 25°C. Miscible with alcohol, chloroform, ether, carbon disulfide, acetone, oils, carbon tetrachloride and glacial acetic acid.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Seiji. Shinkai, et al. Hexasulfonated calix[6]arene derivatives: a new class of catalysts, surfactants, and host molecules.J. Am. Chem. Soc.,1986108(9), 2409-2416.
  2. Seiji Shinkai, et al. New water-soluble host molecules derived from calix[6]arene.Tetrahedron Lett.,198425(46), 5315-5318.