1-Bromo-6-chlorohexane, 97%
1-Bromo-6-chlorohexane, 97%
1-Bromo-6-chlorohexane, 97%
Thermo Scientific Chemicals

1-Bromo-6-chlorohexane, 97%

CAS: 6294-17-3 | C6H12BrCl | 199.52 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B21619.06
also known as B21619-06
Price (EUR)
32,30
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
32,30
Each
Chemical Identifiers
CAS6294-17-3
IUPAC Name1-bromo-6-chlorohexane
Molecular FormulaC6H12BrCl
InChI KeyJTYUIAOHIYZBPB-UHFFFAOYSA-N
SMILESClCCCCCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.4795-1.4845 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only