2,7-Dimethyloctane, 98%, Thermo Scientific Chemicals
2,7-Dimethyloctane, 98%, Thermo Scientific Chemicals
2,7-Dimethyloctane, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,7-Dimethyloctane, 98%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
B21646.03
also known as B21646-03
1 g
Catalog number B21646.03
also known as B21646-03
Price (EUR)
40,50
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
40,50
Each
Specifications
CAS1072-16-8
Chemical Name or Material2,7-Dimethyloctane
DOT InformationTransport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
Density0.72
Formula Weight142.29
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only