2-n-Pentylpropane-1,3-diol, 97%, Thermo Scientific Chemicals
2-n-Pentylpropane-1,3-diol, 97%, Thermo Scientific Chemicals
2-n-Pentylpropane-1,3-diol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-n-Pentylpropane-1,3-diol, 97%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
B21800.06
also known as B21800-06
5 g
Catalog number B21800.06
also known as B21800-06
Price (EUR)
93,30
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
93,30
Each
Chemical Identifiers
CAS25462-23-1
IUPAC Name2-pentylpropane-1,3-diol
Molecular FormulaC8H18O2
InChI KeyRHYUFGNCUXTFTC-UHFFFAOYSA-N
SMILESCCCCCC(CO)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4500-1.4550 @ 20?C
FormLiquid
Appearance (Color)Clear colorless
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only