9-(Trimethylsilyl)fluorene, 99%
9-(Trimethylsilyl)fluorene, 99%
9-(Trimethylsilyl)fluorene, 99%
Thermo Scientific Chemicals

9-(Trimethylsilyl)fluorene, 99%

CAS: 7385-10-6 | C16H18Si | 238.41 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B21816.03
also known as B21816-03
Price (EUR)
122,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
122,00
Each
Chemical Identifiers
CAS7385-10-6
IUPAC Name(9H-fluoren-9-yl)trimethylsilane
Molecular FormulaC16H18Si
InChI KeyYFPRGLHBYSNHMW-UHFFFAOYSA-N
SMILESC[Si](C)(C)C1C2=CC=CC=C2C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (GC)≥98.5%
Melting Point (clear melt)96.0-100.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only