2-Bromobiphenyl, 98%
2-Bromobiphenyl, 98%
2-Bromobiphenyl, 98%
Thermo Scientific Chemicals

2-Bromobiphenyl, 98%

CAS: 2052-07-5 | C12H9Br | 233.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number B21840.14
also known as B21840-14
Price (EUR)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS2052-07-5
IUPAC Name2-bromo-1,1'-biphenyl
Molecular FormulaC12H9Br
InChI KeyKTADSLDAUJLZGL-UHFFFAOYSA-N
SMILESBrC1=CC=CC=C1C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colorless to yellow
FormLiquid
Assay (GC)>97.5%
It is used as an intermediate of organic synthesis and pharmaceutical. It also can react with norborn-2-ene to get 5-phenyl-1,2,3,4,4a,8b-hexahydro-1,4-methanobiphenylene, in synthesis of a novel trispirocyclic hydrocarbon. It is used as a reference compound to investigate extraction efficiency of solid-phase micro extraction fibers, based on methacrylic acid-trimethylolpropanetrimethacrylate copolymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an intermediate of organic synthesis and pharmaceutical. It also can react with norborn-2-ene to get 5-phenyl-1,2,3,4,4a,8b-hexahydro-1,4-methanobiphenylene, in synthesis of a novel trispirocyclic hydrocarbon. It is used as a reference compound to investigate extraction efficiency of solid-phase micro extraction fibers, based on methacrylic acid-trimethylolpropanetrimethacrylate copolymers.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Jingbin Zeng; Jinmei Chen; Liangbi Chen; Yiru Wang; Wenfeng Chen; Xiaoli Huang; Xi Chen. The extraction performance of methacrylic acid-trimethylolpropanetrimethacrylate solid-phase microextraction fibers in aqueous solutions. Analytica Chimica Acta. 2009, 648 (2), 194-199.
  2. Francis H. Case. The Nitration of Certain Halobiphenyls. III. Nitro Derivatives of 2-Chloro- and 2-Bromobiphenyl. J. Am. Chem. Soc. 1943, 65 (11), 2137-2140.