N-Acetylglycine, 99%
N-Acetylglycine, 99%
N-Acetylglycine, 99%
Thermo Scientific Chemicals

N-Acetylglycine, 99%

CAS: 543-24-8 | C4H7NO3 | 117.104 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number B21887.22
also known as B21887-22
Price (EUR)
32,70
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
32,70
Each
Chemical Identifiers
CAS543-24-8
IUPAC Name2-acetamidoacetic acid
Molecular FormulaC4H7NO3
InChI KeyOKJIRPAQVSHGFK-UHFFFAOYSA-N
SMILESCC(=O)NCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥98.5 to ≤102.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only