Digoxin, 96%
Digoxin, 96%
Digoxin, 96%
Thermo Scientific Chemicals

Digoxin, 96%

CAS: 20830-75-5 | C41H64O14 | 780.949 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B21902.03
also known as B21902-03
Price (EUR)
104,65
Online exclusive
116,00
Save 11,35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
104,65
Online exclusive
116,00
Save 11,35 (10%)
Each
Chemical Identifiers
CAS20830-75-5
IUPAC Name4-[(1R,3aS,3bR,5aR,7S,9aS,9bS,11R,11aS)-7-{[(2R,4S,5S,6R)-5-{[(2S,4S,5S,6R)-5-{[(2S,4S,5S,6R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy}-4-hydroxy-6-methyloxan-2-yl]oxy}-4-hydroxy-6-methyloxan-2-yl]oxy}-3a,11-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]-2,5-dihydrofuran-2-one
Molecular FormulaC41H64O14
InChI KeyLTMHDMANZUZIPE-PUGKRICDSA-N
SMILESC[C@H]1O[C@H](C[C@H](O)[C@@H]1O)O[C@H]1[C@@H](O)C[C@H](O[C@H]2[C@@H](O)C[C@H](O[C@H]3CC[C@@]4(C)[C@H](CC[C@@H]5[C@@H]4C[C@@H](O)[C@]4(C)[C@H](CC[C@]54O)C4=CC(=O)OC4)C3)O[C@@H]2C)O[C@@H]1C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥95.0%
Optical Rotation12.0 ± 0.1? (C=10 in pyridine)
FormPowder
Appearance (Color)White
Digoxin is used in several cardiac pathologies such as atrial fibrillation and heart failure.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Digoxin is used in several cardiac pathologies such as atrial fibrillation and heart failure.

Solubility
Soluble in dimethyl sulfoxide and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Barry F. Uretsky et.al. Randomized study assessing the effect of digoxin withdrawal in patients with mild to moderate chronic congestive heart failure: Results of the Proved trial. Journal of the American College of Cardiology., 1993, 22 (4), 955-962.
  2. Rodney H. Falk, M.D.; Anne A. Knowlton, M.D.; Sheilah A. Bernard, M.D.; Norman E. Gotlieb, M.D.; And Nancy J. Battinelli, R.N. Digoxin for Converting Recent-Onset Atrial Fibrillation to Sinus Rhythm: A Randomized, Double-Blinded Trial. Ann Intern Med, 1987, 106 (4), 503-506.