Dimethyl pimelate, 98+%
Dimethyl pimelate, 98+%
Dimethyl pimelate, 98+%
Thermo Scientific Chemicals

Dimethyl pimelate, 98+%

CAS: 1732-08-7 | C9H16O4 | 188.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B22026.22
also known as B22026-22
Price (EUR)
368,00
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
368,00
Each
Chemical Identifiers
CAS1732-08-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Identification (FTIR)Conforms
Refractive Index1.4290-1.4330 @ 20°C
Assay (GC)≥98.5%
Dimethyl pimelate was used as the internal standard in the analysis of phthalates, sebacates, epoxidized soybean oil (ESBO) and polyadipates after transesterification to the ethyl esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl pimelate was used as the internal standard in the analysis of phthalates, sebacates, epoxidized soybean oil (ESBO) and polyadipates after transesterification to the ethyl esters.

Solubility
Soluble in ethanol, benzene, ether. Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Koni Grob. Verification of results to improve the quality of analytical data: a proposal. Journal of Chromatography. 2007, 1150 (1-2), 93-99.
  2. A. T. James and J. Webb. The determination of the structure of unsaturated fatty acids on a micro scale with the gas-liquid chromatogram. Biochem J.. 1957, 66 (3), 515-520.