Thermo Scientific Chemicals

Cobalt(II) chloride, anhydrous, 97%, Thermo Scientific Chemicals

Catalog number: B22031.22
100 g, Each
Thermo Scientific Chemicals

Cobalt(II) chloride, anhydrous, 97%, Thermo Scientific Chemicals

Catalog number: B22031.22
100 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: B22031.22
also known as B22031-22
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
7646-79-9
IUPAC Name
λ²-cobalt(2+) dichloride
Molecular Formula
Cl2Co
InChI Key
GVPFVAHMJGGAJG-UHFFFAOYSA-L
SMILES
[Cl-].[Cl-].[Co++]
Form
powder
Comment
Ni <0.2%
Appearance (Color)
Blue
Assay (unspecified)
>96.0% Based on CoCl2 Content
Comment
Pb <0.01%

Description

Cobalt(II) chloride is used in humidity indicator in weather instruments. In the anhydrous form, it finds use in electroplating of cobalt, in organic chemistry and is a precursor to cobaltocene, (bis(cyclopentadienyl)cobalt(II), which is a good reducing agent. It also serves as a Lewis acid. Cobalt chloride is an indicator for water in desiccants, owing to the reversible hydration/dehydration coupled with distinct color change. Cobalt chloride is useful for producing invisible ink as it turns blue when heated and becomes invisible once it gets cooled. Cobalt(II) chloride catalyzes cross coupling of aryl halides or vinyl halides with aryl grignard reagents in excellent yields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cobalt(II) chloride is used in humidity indicator in weather instruments. In the anhydrous form, it finds use in electroplating of cobalt, in organic chemistry and is a precursor to cobaltocene, (bis(cyclopentadienyl)cobalt(II), which is a good reducing agent. It also serves as a Lewis acid. Cobalt chloride is an indicator for water in desiccants, owing to the reversible hydration/dehydration coupled with distinct color change. Cobalt chloride is useful for producing invisible ink as it turns blue when heated and becomes invisible once it gets cooled. Cobalt(II) chloride catalyzes cross coupling of aryl halides or vinyl halides with aryl grignard reagents in excellent yields.

Solubility
Freely soluble in water and methanol. Soluble in alcohol, acetone, ether, glycerol, and pyridine.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 4
    Acute Inhalation Toxicity - Dusts and Mists
    Category 4
    Serious Eye Damage/Eye Irritation
    Category 1
    Respiratory Sensitization
    Category 1
    Skin Sensitization
    Category 1
    Germ Cell Mutagenicity
    Category 2
    Carcinogenicity
    Category 1B
    Reproductive Toxicity
    Category 1B
    Acute aquatic toxicity
    Category 1
    Chronic aquatic toxicity
    Category 1
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H302 + H332 - Harmful if swallowed or if inhaled

    H317 - May cause an allergic skin reaction

    H318 - Causes serious eye damage

    H334 - May cause allergy or asthma symptoms or breathing difficulties if inhaled

    H341 - Suspected of causing genetic defects

    H350i - May cause cancer by inhalation

    H360F - May damage fertility

    H410 - Very toxic to aquatic life with long lasting effects

    Precautionary Statements

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

    P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

    P310 - Immediately call a POISON CENTER or doctor/physician

    Additional EU labelling

    Restricted to professional users