Sodium triacetoxyborohydride, 95%
Sodium triacetoxyborohydride, 95%
Sodium triacetoxyborohydride, 95%
Thermo Scientific Chemicals

Sodium triacetoxyborohydride, 95%

CAS: 56553-60-7 | C6H10BNaO6 | 211.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number B22060.22
also known as B22060-22
Price (EUR)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS56553-60-7
IUPAC Namesodium bis(acetyloxy)boranuidyl acetate
Molecular FormulaC6H10BNaO6
InChI KeyHHYFEYBWNZJVFQ-UHFFFAOYSA-N
SMILES[Na+].CC(=O)O[BH-](OC(C)=O)OC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay from Supplier's CofA≥94.0%
FormPowder
Sodium triacetoxyborohydride is used as a reagent in reductive amination of ketones, aldehydes and lactamization of carbonyl compounds with amines. It also serves as a reducing agent in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium triacetoxyborohydride is used as a reagent in reductive amination of ketones, aldehydes and lactamization of carbonyl compounds with amines. It also serves as a reducing agent in organic synthesis.

Solubility
Soluble in dimethyl sulfoxide, methanol, benzene, toluene, terahydrofuran, dioxane and methylene chloride.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, water, alcohols and acids.
RUO – Research Use Only

General References:

  1. Selective reducing agent; review: Org. Prep. Proced. Int., 17, 317 (1985). Reduces aldehydes in the presence of ketones: J. Chem. Soc., Chem. Commun., 535 (1975). For stereoselective reduction of ketones in AcOH, see: Tetrahedron Lett., 273 (1983). Reduces ß-hydroxy ketones diastereoselectively to anti-diols: J. Am. Chem. Soc., 110, 3560 (1988), and cyclic ß-enamino esters selectively to the cis-amino esters: J. Org. Chem., 59, 5328 (1994); Tetrahedron: Asymmetry, 5, 1455 (1994).
  2. Excellent reagent for reductive amination of aldehydes and ketones with a wide variety of amines: J. Org. Chem., 61, 3849 (1996); Tetrahedron Lett., 37, 3977 (1996); review: A.C.S. Symp. Ser., 641, 201 (1996).
  3. Tahara, Y. K.; Ito, M.; Kanyiva, K. S.; Shibata, T. Total Synthesis of cis-Clavicipitic Acid from Asparagine via Ir-Catalyzed C-H bond Activation as a Key Step. Chem. Eur. J. 2015, 21 (32), 11340-11343.
  4. Kang, B.; Oh, J. A.; Lee, J. Y.; Rhim, H.; Yune, T. Y.; Choo, H. Y. P. 3-Benzamides and 3, 4, 5-trimethoxyphenyl amines as calcium channel blockers. Bioorg. Med. Chem 2015, 23 (18), 6166-6172.