Thermo Scientific Chemicals

Amberchrom™ 50WX8 50-100 (H), Thermo Scientific Chemicals

Catalog number: B22109.0E
2500 g, Each
Thermo Scientific Chemicals

Amberchrom™ 50WX8 50-100 (H), Thermo Scientific Chemicals

Catalog number: B22109.0E
2500 g, Each
Quantity
Catalog number: B22109.0E
also known as B22109-0E
Price (EUR)
Price: 1.201,00
Your price:
Quantity
-

Chemical Identifiers

CAS
11119-67-8
MDL Number
MFCD00132726
Appearance (Color)
White to pale brown
Form
Beads
Comment
Ionic form: H+
Comment
Sulfonic acid active functional group
Exchange capacity
≥1.7 eq/L (H)

Description

Dowex™ 50WX8 50-100 (H) is used in deionizing supernatant fraction samples for paper chromatography and as a catalyst. It is involved in the separation medium for thin layer chromatography, paper chromatography and low pressure liquid chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dowex|r 50WX8 50-100 (H) is used in deionizing supernatant fraction samples for paper chromatography and as a catalyst. It is involved in the separation medium for thin layer chromatography, paper chromatography and low pressure liquid chromatography.

Solubility
Insoluble in water.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Serious Eye Damage/Eye Irritation
    Category 1
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H318 - Causes serious eye damage

    Precautionary Statements

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

    P310 - Immediately call a POISON CENTER or doctor/physician