5-Amino-2,4,6-triiodoisophthalic acid, 95%
5-Amino-2,4,6-triiodoisophthalic acid, 95%
5-Amino-2,4,6-triiodoisophthalic acid, 95%
Thermo Scientific Chemicals

5-Amino-2,4,6-triiodoisophthalic acid, 95%

CAS: 35453-19-1 | C8H4I3NO4 | 558.836 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number B22178.09
also known as B22178-09
Price (EUR)
26,90
Each
Quantity:
10 g
Request bulk or custom format
Price (EUR)
26,90
Each
Chemical Identifiers
CAS35453-19-1
IUPAC Name5-amino-2,4,6-triiodobenzene-1,3-dicarboxylate
Molecular FormulaC8H2I3NO4
InChI KeyJEZJSNULLBSYHV-UHFFFAOYSA-L
SMILESNC1=C(I)C(C([O-])=O)=C(I)C(C([O-])=O)=C1I
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥94.0%
Assay (Aqueous acid-base Titration)≥94.0 to ≤106.0%
Appearance (Color)Cream to pale brown
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only