4-Chloro-2-methyl-6-nitroaniline, 98%
4-Chloro-2-methyl-6-nitroaniline, 98%
4-Chloro-2-methyl-6-nitroaniline, 98%
4-Chloro-2-methyl-6-nitroaniline, 98%
Thermo Scientific Chemicals

4-Chloro-2-methyl-6-nitroaniline, 98%

CAS: 62790-50-5 | C7H7ClN2O2 | 186.60 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B22253.03
also known as B22253-03
Price (EUR)
18,90
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (EUR)
18,90
Each
Add to cart
Chemical Identifiers
CAS62790-50-5
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Appearance (Color)Orange
Melting Point (clear melt)126.0-132.0?C
FormCrystals or powder or crystalline powder or needles
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only