2,2'-Thiodiethanethiol, tech. 85%
2,2'-Thiodiethanethiol, tech. 85%
2,2'-Thiodiethanethiol, tech. 85%
Thermo Scientific Chemicals

2,2'-Thiodiethanethiol, tech. 85%

CAS: 3570-55-6 | C4H10S3 | 154.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number B22304.09
also known as B22304-09
Price (EUR)
73,10
Each
Quantity:
10 g
Request bulk or custom format
Price (EUR)
73,10
Each
Chemical Identifiers
CAS3570-55-6
IUPAC Name2-[(2-sulfanylethyl)sulfanyl]ethane-1-thiol
Molecular FormulaC4H10S3
InChI KeyKSJBMDCFYZKAFH-UHFFFAOYSA-N
SMILESSCCSCCS
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥80.0%
Refractive Index1.5555-1.6355 @ 20°C
Appearance (Color)Clear colorless
2,2?-Thiodiethanol is used in the synthesis of novel luminescent polynuclear gold(I) phosphine complexes 2,2'-Thiodiethanethiol is used for extraction photometric determination of Ni, gives color reaction with metals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2′-Thiodiethanol is used in the synthesis of novel luminescent polynuclear gold(I) phosphine complexes 2,2′-Thiodiethanethiol is used for extraction photometric determination of Ni, gives color reaction with metals.

Solubility
Difficult to mix.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Steven G. O'Neal.; Steven R. Earle.; Ronald R. Fisher. The effect of metal ions on mitochondrial pyridine dinucleotide transhydrogenase. Biochimica et Biophysica Acta (BBA) - Bioenergetics. 1980, 589, (2), 217-230.
  2. Shubhamoy Chowdhury.; Nobuyoshi Koshino.; Ali Canlier.; Koichiro Mizuoka.; Yasuhisa Ikeda. Molecular structure and linkage isomerization of isothiocyanato(3-thiapentane-1,5-dithiolato)oxorhenium(V) complex. Inorganica Chimica Acta. 2006, 359, (8), 2472-2478.