trans-4-Aminocyclohexanol, 98+%
trans-4-Aminocyclohexanol, 98+%
trans-4-Aminocyclohexanol, 98+%
trans-4-Aminocyclohexanol, 98+%
Thermo Scientific Chemicals

trans-4-Aminocyclohexanol, 98+%

CAS: 27489-62-9 | C6H13NO | 115.176 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B22365.22
also known as B22365-22
Price (EUR)
387,00
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (EUR)
387,00
Each
Add to cart
Chemical Identifiers
CAS27489-62-9
IUPAC Name4-aminocyclohexan-1-ol
Molecular FormulaC6H13NO
InChI KeyIMLXLGZJLAOKJN-UHFFFAOYSA-N
SMILESNC1CCC(O)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.0%
Appearance (Color)White to pale cream to pale brown
FormCrystals or powder or crystalline powder or fused solid
Melting Point (clear melt)108.0-115.0?C
trans-4-Aminocyclohexanol is used as raw material in organic synthesis and is an important intermediate in the synthesis of drugs such as Ambroxol hydrochloride. It can react with butyric acid-(2-chloro-ethyl ester) to get butyric acid 4-amino-cyclohexyl ester. This reaction needs catalytic agent Aspergillus niger lipase and solvent 2-methyl-butan-2-ol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-4-Aminocyclohexanol is used as raw material in organic synthesis and is an important intermediate in the synthesis of drugs such as Ambroxol hydrochloride. It can react with butyric acid-(2-chloro-ethyl ester) to get butyric acid 4-amino-cyclohexyl ester. This reaction needs catalytic agent Aspergillus niger lipase and solvent 2-methyl-butan-2-ol.

Solubility
Soluble in water.

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Thomas P. Johnston.; George S. McCaleb.; John A. Montgomery. Synthesis and biologic evaluation of major metabolites of N-(2-chloroethyl)-N'-cyclohexyl-N-nitrosourea. J. Med. Chem. 1975, 18, (6), 634-637.
  2. Jie Cheng.; Mark L. Trudell. Synthesis of N-Heteroaryl-7-azabicyclo[2.2.1]heptane Derivatives via Palladium-Bisimidazol-2-ylidene Complex Catalyzed Amination Reactions. Org. Lett.2001, 3, (9), 1371-1374.