2-Methyl-6-nitrobenzoxazole, 98%
2-Methyl-6-nitrobenzoxazole, 98%
2-Methyl-6-nitrobenzoxazole, 98%
Thermo Scientific Chemicals

2-Methyl-6-nitrobenzoxazole, 98%

CAS: 5683-43-2 | C8H6N2O3 | 178.147 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B22415.14
also known as B22415-14
Price (EUR)
314,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
314,00
Each
Chemical Identifiers
CAS5683-43-2
IUPAC Name2-methyl-6-nitro-1,3-benzoxazole
Molecular FormulaC8H6N2O3
InChI KeyDPVZLXWUFYMLBK-UHFFFAOYSA-N
SMILESCC1=NC2=CC=C(C=C2O1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale brown
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Melting Point (clear melt)153.0-159.0?C
2-Methyl-6-nitrobenzoxazole is used as a flavoring agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methyl-6-nitrobenzoxazole is used as a flavoring agents.

Solubility
Soluble in oils. Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
Daniel Best.; Szymon Kujawa.; Hon Wai Lam. Diastereo- and Enantioselective Pd(II)-Catalyzed Additions of 2-Alkylazaarenes to N-Boc Imines and Nitroalkenes. J. Am. Chem. Soc. 2012, 134, (44), 18193-18196.