7-Oxabicyclo[2.2.1]heptane, 98%
7-Oxabicyclo[2.2.1]heptane, 98%
7-Oxabicyclo[2.2.1]heptane, 98%
Thermo Scientific Chemicals

7-Oxabicyclo[2.2.1]heptane, 98%

CAS: 279-49-2 | C6H10O | 98.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B22612.06
also known as B22612-06
Price (EUR)
83,70
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
83,70
Each
Chemical Identifiers
CAS279-49-2
IUPAC Name7-oxabicyclo[2.2.1]heptane
Molecular FormulaC6H10O
InChI KeyYPWFNLSXQIGJCK-UHFFFAOYSA-N
SMILESC1CC2CCC1O2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
Refractive Index1.4460-1.4500 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only