Benzo[b]thiophene-2-boronic acid, 97%
Benzo[b]thiophene-2-boronic acid, 97%
Benzo[b]thiophene-2-boronic acid, 97%
Thermo Scientific Chemicals

Benzo[b]thiophene-2-boronic acid, 97%

CAS: 98437-23-1 | C8H7BO2S | 178.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B22835.03
also known as B22835-03
Price (EUR)
35,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
35,00
Each
Chemical Identifiers
CAS98437-23-1
IUPAC Name(1-benzothiophen-2-yl)boronic acid
Molecular FormulaC8H7BO2S
InChI KeyYNCYPMUJDDXIRH-UHFFFAOYSA-N
SMILESOB(O)C1=CC2=CC=CC=C2S1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Assay (Aqueous acid-base Titration)≥96.0%
FormPowder
Proton NMRConforms to structure
Benzo[b]thiophene-2-boronic acid is used in the synthesis of phosphorescent sensor for quantification of copper(II) ion, UV promoted phenanthridine syntheses, Preparation of CYP11B1 inhibitors for treatment of cortisol dependent diseases, PDE4 inhibitors, Suzuki-Miyaura cross-coupling reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Insoluble in water.

Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Frédéric J.J. Bihel; Hélène Justiniano; Martine Schmitt; Malik Hellal; Mohamed A. Ibrahim; Claire Lugnier; Jean-Jacques Bourguigno. New PDE4 inhibitors based on pharmacophoric similarity between papaverine and tofisopam. Bioorganic & Medicinal Chemistry Letters. 2011, 21 (21), 6567-6572.
  2. Partha S. Banerjee; Edison S. Zuniga; Iwao Ojima; Isaac S. Carrico. Targeted and armed oncolytic adenovirus via chemoselective modification. Bioorganic & Medicinal Chemistry Letters. 2011, 21, (17), 4985-4988.