Cystamine dihydrochloride, 97+%
Cystamine dihydrochloride, 97+%
Cystamine dihydrochloride, 97+%
Thermo Scientific Chemicals

Cystamine dihydrochloride, 97+%

CAS: 56-17-7 | C4H14Cl2N2S2 | 225.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number B22873.14
Price (EUR)
39,90
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
39,90
Each
Chemical Identifiers
CAS56-17-7
IUPAC Namedihydrogen 2-[(2-aminoethyl)disulfanyl]ethan-1-amine dichloride
Molecular FormulaC4H14Cl2N2S2
InChI KeyYUFRRMZSSPQMOS-UHFFFAOYSA-N
SMILES[H+].[H+].[Cl-].[Cl-].NCCSSCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Water Content (Karl Fischer Titration)≤1.5%
FormPowder
Assay (Titration ex Chloride)≥97.0 to ≤103.0%
Melting Point (clear melt)213.0-223.0?C
Appearance (Color)White to cream
Cystamine dihydrochloride acts as an anti-infective agent, which is used in the treatment of urinary tract infections. It is also used as a radiation-protective agent that interferes with sulfhydryl enzymes. Further, it serves as a heparin antagonist and sulfhydryl modifying reagent. In addition to this, it is used as an inhibitor of TGase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cystamine dihydrochloride acts as an anti-infective agent, which is used in the treatment of urinary tract infections. It is also used as a radiation-protective agent that interferes with sulfhydryl enzymes. Further, it serves as a heparin antagonist and sulfhydryl modifying reagent. In addition to this, it is used as an inhibitor of TGase.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

General References:

  1. Shi, H.; Zhou, K.; Yu, Q.; Cui, Z.; Jiang, Y.; Lu, X.; Cai, Y. Programmable self-assembly of a cystamine-block copolymer in response to pH and progressive reduction-ionization-oxidation. Polym. Chem. 2015, 6 (42), 7455-7463.
  2. Li, H.; Jiang, H.; Zhao, M.; Fu, Y.; Sun, X. Intracellular redox potential-responsive micelles based on polyethylenimine-cystamine-poly(epsilon-caprolactone) block copolymer for enhanced miR-34a delivery. Poly. Chem. 2015, 6 (11), 1952-1960.